Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ldlr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252658.1 | NP_001239587.1 | 7, 8 | 1265 | ||
NM_001252659.1 | NP_001239588.1 | 7, 8 | 1262 | |||
NM_010700.3 | NP_034830.2 | 7, 8 | 1265 | |||
GenBank mRNA | AF425607.1 | - | 7, 8 | 1113 | ||
AK076243.1 | - | 7, 8 | 1243 | |||
AK149950.1 | - | 7, 8 | 1203 | |||
AK151968.1 | - | 7, 8 | 1232 | |||
AK152103.1 | - | 7, 8 | 1201 | |||
AK160006.1 | - | 7, 8 | 1203 | |||
AK161104.1 | - | 7, 8 | 1203 | |||
AK166466.1 | - | 7, 8 | 1202 | |||
AK170264.1 | - | 7, 8 | 1210 | |||
BC019207.1 | - | 7, 8 | 1183 | 5052599 | ||
BC053041.1 | - | 7, 8 | 1210 | 5704871 | ||
X64414.1 | - | 7, 8 | 1184 | |||
Z19521.1 | - | 7, 8 | 1063 |