Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lmnb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010722.5 | NP_034852.2 | 5 | 776 | ||
GenBank mRNA | AK132671.1 | - | 2 | 484 | ||
BC042430.1 | - | 5 | 781 | 3674002 | ||
BC051985.1 | - | 5 | 752 | 5695459 | ||
D13455.1 | - | 2 | 463 | |||
X54098.1 | - | 5 | 735 |