Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR155 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033045.3 | NP_001028217.1 | 3 | 582 | ||
NM_001267050.1 | NP_001253979.1 | 3 | 479 | |||
NM_001267051.1 | NP_001253980.1 | 2 | 433 | |||
NM_152529.6 | NP_689742.4 | 2 | 433 | |||
XM_017003485.1 | XP_016858974.1 | 4 | 624 | |||
XM_017003486.1 | XP_016858975.1 | 2 | 485 | |||
XM_017003487.1 | XP_016858976.1 | 3 | 486 | |||
GenBank mRNA | AK091200.1 | - | 2 | 374 | ||
AK307896.1 | - | 2 | 425 | |||
AK315125.1 | - | 2 | 374 | |||
AL834154.1 | - | 3 | 531 | |||
BC028730.1 | - | 3 | 528 | 4813561 | ||
BC035037.1 | - | 3 | 490 | 5260361 | ||
BC036681.1 | - | 2 | 379 | |||
BI548910.1 | - | 3 | 490 | |||
BX537947.1 | - | 2 | 426 | |||
DC334438.1 | - | 2 | 433 |