Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Marco | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010766.2 | NP_034896.1 | 15 | 1545 | ||
GenBank mRNA | AK156388.1 | - | 15 | 1219 | ||
AK159094.1 | - | 2 | 50 | |||
AK159967.1 | - | 15 | 1227 | |||
AK169230.1 | - | 15 | 1221 | |||
BC132338.1 | - | 15 | 1485 | 40130615 | ||
BC137939.1 | - | 15 | 1485 | 8860958 | ||
U18424.1 | - | 15 | 1397 |