Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mbd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010774.2 | NP_034904.2 | 4 | 1296 | ||
GenBank mRNA | AF072249.1 | - | 4 | 1313 | ||
AK161005.1 | - | 4 | 2283 | |||
BC024812.1 | - | 4 | 1278 | 5356016 |