Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mbl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010775.2 | NP_034905.1 | 4 | 439 | ||
GenBank mRNA | AK034788.1 | - | 4 | 439 | ||
BC012245.1 | - | 4 | 401 | 4193682 | ||
BC021762.1 | - | 4 | 393 | 5132514 | ||
D11441.1 | - | 4 | 383 | |||
S42292.1 | - | 4 | 439 |