Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mdfi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109973.2 | NP_001103443.1 | 5 | 1488 | ||
NM_001276390.1 | NP_001263319.1 | 4 | 1370 | |||
NM_001276391.1 | NP_001263320.1 | 6 | 1260 | |||
NM_010783.3 | NP_034913.2 | 5 | 1156 | |||
GenBank mRNA | BC006018.1 | - | 4 | 1123 | 3488088 | |
BC010259.1 | - | 5 | 1328 | 3481922 | ||
BC083058.1 | - | 5 | 1261 | 5354539 | ||
BC085233.1 | - | 5 | 1488 | 5318215 | ||
U57820.1 | - | 4 | 1128 | |||
U57822.1 | - | 4 | 858 | |||
W65565.1 | - | 1 | 373 |