Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mid1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290504.1 | NP_001277433.1 | 677 | |||
NM_001290505.1 | NP_001277434.1 | 901 | ||||
NM_010797.3 | NP_034927.2 | 784 | ||||
GenBank mRNA | AF026565.1 | - | 677 | |||
AK135734.1 | - | 900 | ||||
AY540038.1 | - | 89 | ||||
AY540040.1 | - | 89 | ||||
AY540042.1 | - | 89 | ||||
Y14848.1 | - | 738 |