Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mlf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039543.2 | NP_001034632.1 | 8 | 947 | ||
NM_010801.3 | NP_034931.2 | 7 | 902 | |||
GenBank mRNA | AF009515.1 | - | 7 | 843 | ||
AF100171.1 | - | 7 | 843 | |||
AK133448.1 | - | 7 | 903 | |||
BC027526.1 | - | 8 | 896 | 1495721 |