Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd200 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010818.3 | NP_034948.3 | 3 | 365 | ||
GenBank mRNA | AF004023.1 | - | 3 | 203 | ||
AK148308.1 | - | 3 | 334 | |||
AK164478.1 | - | 3 | 288 | |||
BC019172.1 | - | 3 | 235 | 5003655 | ||
BC051984.1 | - | 3 | 283 | 5707683 | ||
BC054759.1 | - | 3 | 298 | 6413363 | ||
CN665640.1 | - | 3 | 365 |