Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038609.2 | NP_001033698.1 | 2 | 269 | ||
NM_001285454.1 | NP_001272383.1 | 2 | 269 | |||
NM_001285455.1 | NP_001272384.1 | 2 | 269 | |||
NM_001285456.1 | NP_001272385.1 | 2 | 269 | |||
NM_010838.4 | NP_034968.3 | 2 | 269 | |||
GenBank mRNA | AF483518.1 | - | 1 | 84 | ||
AF483519.1 | - | 1 | 84 | |||
AK038797.1 | - | 2 | 226 | |||
AK078167.1 | - | 2 | 243 | |||
AK147636.1 | - | 2 | 230 | |||
BC014748.1 | - | 2 | 207 | 4504850 | ||
BY249380.1 | - | 2 | 264 | |||
CO432693.1 | - | 2 | 219 | |||
M18775.1 | - | 2 | 270 | |||
M18776.1 | - | 2 | 270 | |||
U12914.1 | - | 2 | 153 | |||
U12915.1 | - | 2 | 153 | |||
U12916.1 | - | 2 | 153 | |||
W18770.1 | - | 2 | 251 |