Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039657.2 | NP_001034746.1 | 4, 5 | 839 | ||
NM_001039658.2 | NP_001034747.1 | 4, 5 | 839 | |||
NM_001286557.1 | NP_001273486.1 | 5, 6 | 923 | |||
NM_001286558.1 | NP_001273487.1 | 4, 5 | 854 | |||
GenBank mRNA | AB057423.1 | - | 4, 5 | 827 | ||
AF329359.1 | - | 4, 5 | 864 | |||
AF329360.1 | - | 5, 6 | 933 | |||
AK015724.1 | - | 4, 5 | 780 | |||
AK015732.1 | - | 4, 5 | 812 | |||
AK161511.1 | - | 4, 5 | 840 | |||
BC024377.1 | - | 4, 5 | 815 | 4976492 | ||
U67176.1 | - | 6, 7 | 856 | |||
U77383.1 | - | 4, 5 | 476 |