Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nkx2-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010919.2 | NP_035049.1 | 2 | 1449 | ||
GenBank mRNA | AK141002.1 | - | 2 | 1454 | ||
BC138159.1 | - | 3 | 1270 | 8861178 | ||
BC138160.1 | - | 3 | 1270 | 8861179 | ||
U31566.1 | - | 2 | 1408 |