Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nnat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291128.1 | NP_001278057.1 | 3 | 847 | ||
NM_001291129.1 | NP_001278058.1 | 3 | 835 | |||
NM_001291130.1 | NP_001278059.1 | 3 | 786 | |||
NM_010923.3 | NP_035053.1 | 3 | 840 | |||
NM_180960.3 | NP_851291.1 | 2 | 759 | |||
GenBank mRNA | AB004048.1 | - | 3 | 1046 | ||
AK003004.1 | - | 3 | 804 | |||
AK160626.1 | - | 3 | 808 | |||
BC036984.1 | - | 3 | 798 | 4981517 | ||
BU705167.1 | - | 1 | 101 | |||
CJ302750.1 | - | 1 | 14 | |||
GQ184462.1 | - | 3 | 742 | |||
X83568.1 | - | 3 | 608 | |||
X83569.1 | - | 2 | 715 | |||
X83570.1 | - | 3 | 796 |