Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rrp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010925.2 | NP_035055.2 | 1, 2 | 232 | ||
GenBank mRNA | AK077540.1 | - | 1, 2 | 157 | ||
BC065993.1 | - | 1, 2 | 194 | 5684672 | ||
BC065994.1 | - | 1, 2 | 152 | 5685269 | ||
BY106122.1 | - | 1, 2 | 232 | |||
U79773.1 | - | 1, 2 | 184 | |||
U79774.1 | - | 1, 2 | 184 |