Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uhrf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111078.1 | NP_001104548.1 | 2 | 173 | ||
NM_001111079.1 | NP_001104549.1 | 2 | 317 | |||
NM_001111080.1 | NP_001104550.1 | 2 | 173 | |||
NM_010931.3 | NP_035061.3 | 2 | 317 | |||
GenBank mRNA | AF274046.1 | - | 2 | 230 | ||
AK143688.1 | - | 2 | 142 | |||
AK145376.1 | - | 2 | 139 | |||
AK145419.1 | - | 2 | 138 | |||
AK145543.1 | - | 2 | 257 | |||
AK145583.1 | - | 2 | 139 | |||
AK146951.1 | - | 2 | 290 | |||
AK147046.1 | - | 2 | 257 | |||
AK150489.1 | - | 2 | 173 | |||
AK151701.1 | - | 2 | 139 | |||
AK151837.1 | - | 2 | 173 | |||
AK152930.1 | - | 2 | 173 | |||
AK153083.1 | - | 2 | 173 | |||
BC022167.1 | - | 2 | 245 | 4017286 | ||
D87908.1 | - | 2 | 228 |