Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUP214 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318324.1 | NP_001305253.1 | 2 | 224 | ||
NM_005085.3 | NP_005076.3 | 2 | 224 | |||
XM_005272216.3 | XP_005272273.1 | 2 | 224 | |||
XM_005272218.3 | XP_005272275.1 | 2 | 224 | |||
XM_005272219.3 | XP_005272276.1 | 2 | 224 | |||
XM_006717292.3 | XP_006717355.1 | 2 | 224 | |||
XM_011519053.2 | XP_011517355.1 | 2 | 224 | |||
XM_011519055.2 | XP_011517357.1 | 2 | 224 | |||
XM_017015167.1 | XP_016870656.1 | 2 | 224 | |||
GenBank mRNA | AB159230.1 | - | 2 | 83 | ||
AK307573.1 | - | 2 | 196 | |||
BC012500.1 | - | 2 | 168 | |||
BC045620.1 | - | 2 | 168 | 5268515 | ||
BC105998.1 | - | 2 | 199 | 30531438 | ||
BM455135.1 | - | 191 | ||||
D14689.1 | - | 2 | 135 | |||
DB062584.1 | - | 2 | 226 | |||
X64228.1 | - | 2 | 174 |