Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252250.1 | NP_001239179.1 | 2, 3 | 807 | ||
NM_001252251.1 | NP_001239180.1 | 2, 3 | 807 | |||
NM_001252252.1 | NP_001239181.1 | 2, 3 | 807 | |||
NM_001252253.1 | NP_001239182.1 | 2, 3 | 272 | |||
NM_010898.4 | NP_035028.2 | 2, 3 | 807 | |||
GenBank mRNA | AK017343.1 | - | 2, 3 | 273 | ||
AK045998.1 | - | 2, 3 | 629 | |||
AK180308.1 | - | 1, 2 | 95 | |||
AK185632.1 | - | 1, 2 | 95 | |||
AK213371.1 | - | 1, 2 | 95 | |||
BC005442.1 | - | 2, 3 | 673 | 3496053 | ||
CN715904.1 | - | 2, 3 | 365 | |||
L22987.1 | - | 2, 3 | 265 | |||
L22988.1 | - | 2, 3 | 265 | |||
L22989.1 | - | 2, 3 | 265 | |||
L27090.1 | - | 2, 3 | 258 | |||
L27105.1 | - | 2, 3 | 258 | |||
L28176.1 | - | 2, 3 | 565 | |||
X74671.1 | - | 2, 3 | 824 |