Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010911.2 | NP_035041.2 | 5 | 488 | ||
GenBank mRNA | AJ222660.1 | - | 5 | 487 | ||
AK005724.1 | - | 5 | 477 | |||
AK075575.1 | - | 5 | 486 | |||
AK132838.1 | - | 5 | 489 | |||
AK148527.1 | - | 5 | 489 | |||
AK159828.1 | - | 5 | 477 | |||
BC010586.1 | - | 5 | 466 | 3711352 |