Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nktr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010918.2 | NP_035048.3 | 4 | 333 | ||
GenBank mRNA | AK035582.1 | - | 3 | 234 | ||
AK036649.1 | - | 3 | 1486 | |||
AK088056.1 | - | 4 | 272 | |||
AK144214.1 | - | 4 | 244 | |||
AK163916.1 | - | 4 | 346 | |||
BC048414.1 | - | 3 | 267 | |||
L04289.2 | - | 4 | 338 |