Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nov | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010930.4 | NP_035060.1 | 4 | 735 | ||
GenBank mRNA | AK039481.1 | - | 4 | 726 | ||
AK081310.1 | - | 4 | 753 | |||
AK081944.1 | - | 4 | 961 | |||
AK136407.1 | - | 4 | 755 | |||
AK155048.1 | - | 4 | 754 | |||
BC003774.1 | - | 4 | 741 | 3494896 | ||
X96585.1 | - | 4 | 735 | |||
Y09257.1 | - | 4 | 723 |