Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nsmaf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010945.2 | NP_035075.2 | 2, 3 | 377 | ||
GenBank mRNA | AF013632.1 | - | 2, 3 | 370 | ||
AK039902.1 | - | 2, 3 | 228 | |||
AK049887.1 | - | 2, 3 | 362 | |||
AK134518.1 | - | 2, 3 | 132 | |||
AK147709.1 | - | 2, 3 | 237 | |||
AK164305.1 | - | 2, 3 | 377 | |||
BC066075.1 | - | 2, 3 | 359 | |||
BC082810.1 | - | 2, 3 | 359 | |||
BC145961.1 | - | 2, 3 | 191 | 40131264 | ||
BC145963.1 | - | 2, 3 | 191 | 40131266 | ||
BY729997.1 | - | 2, 3 | 378 |