Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Meis2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136072.2 | NP_001129544.1 | 13 | 2872 | ||
NM_001159567.1 | NP_001153039.1 | 13 | 2893 | |||
NM_001159568.1 | NP_001153040.1 | 12 | 2798 | |||
NM_001159569.1 | NP_001153041.1 | 13 | 3166 | |||
NM_001159570.1 | NP_001153042.1 | 13 | 3145 | |||
NM_010825.3 | NP_034955.1 | 12 | 2777 | |||
GenBank mRNA | AJ000504.1 | - | 13 | 2882 | ||
AJ000505.1 | - | 13 | 2861 | |||
AJ000506.1 | - | 12 | 2787 | |||
AJ000507.1 | - | 12 | 2766 | |||
AK146633.1 | - | 13 | 2872 | |||
BC017375.1 | - | 12 | 2846 | 4191098 | ||
U57343.1 | - | 2929 |