Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048167.1 | NP_001041632.1 | 2, 3 | 2089 | ||
NM_010837.3 | NP_034967.2 | 2, 3 | 2089 | |||
GenBank mRNA | AK018193.2 | - | 2, 3 | 1569 | ||
AK087413.1 | - | 2, 3 | 2052 | |||
BC053039.1 | - | 2, 3 | 2090 | 5709455 |