Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myd88 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010851.2 | NP_034981.1 | 6 | 1638 | ||
GenBank mRNA | AK090107.1 | - | 4 | 2151 | ||
AK150083.1 | - | 6 | 1521 | |||
AK151748.1 | - | 6 | 1594 | |||
AK152596.1 | - | 6 | 1623 | |||
AK152935.1 | - | 6 | 1596 | |||
AK167929.1 | - | 6 | 1619 | |||
AK217564.1 | - | 1 | 367 | |||
BC005591.1 | - | 6 | 1638 | 3482246 | ||
BC051075.1 | - | 6 | 1585 | |||
BC058787.1 | - | 6 | 1573 | 6309465 | ||
X51397.1 | - | 6 | 1507 |