Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myef2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162417.1 | NP_001155889.1 | 15 | 1687 | ||
NM_001162418.1 | NP_001155890.1 | 13 | 1564 | |||
NM_010852.2 | NP_034982.2 | 14 | 1636 | |||
GenBank mRNA | AF483504.1 | - | 13 | 1362 | ||
AF483505.1 | - | 13 | 1362 | |||
AK012133.1 | - | 13 | 1566 | |||
AK048404.1 | - | 15 | 1654 | |||
AK143230.1 | - | 14 | 1572 | |||
U13262.1 | - | 10 | 1077 |
Gene Symbol: Gm9833 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC060946.1 | - | 1 | 1270 | 6741255 |