Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncam1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081445.1 | NP_001074914.1 | 7 | 1175 | ||
NM_001113204.1 | NP_001106675.1 | 7 | 1175 | |||
NM_010875.3 | NP_035005.2 | 7 | 1175 | |||
GenBank mRNA | AK051197.1 | - | 7 | 1032 | ||
AK082621.1 | - | 7 | 1028 | |||
BC011310.1 | - | 7 | 864 | 3708650 | ||
BC062942.1 | - | 7 | 978 | |||
X15049.1 | - | 7 | 996 | |||
Y00051.1 | - | 7 | 996 |