Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfia | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122952.1 | NP_001116424.1 | 11 | 2553 | ||
NM_001122953.1 | NP_001116425.1 | 10 | 2663 | |||
NM_010905.3 | NP_035035.1 | 11 | 2792 | |||
GenBank mRNA | AK052204.1 | - | 11 | 2614 | ||
AK054310.1 | - | 11 | 2661 | |||
AK137731.1 | - | 11 | 2655 | |||
AK139357.1 | - | 11 | 2554 | |||
BC075702.1 | - | 12 | 2725 | 30087137 | ||
D90173.1 | - | 11 | 2631 | |||
Y07690.1 | - | 11 | 2525 | |||
Y07691.1 | - | 10 | 2394 |