Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfix | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081981.2 | NP_001075450.1 | 8 | 2200 | ||
NM_001081982.2 | NP_001075451.1 | 8 | 1525 | |||
NM_001297601.1 | NP_001284530.1 | 7 | 2074 | |||
NM_010906.3 | NP_035036.1 | 7 | 1402 | |||
GenBank mRNA | AK080940.1 | - | 1 | 74 | ||
AK158568.1 | - | 7 | 2077 | |||
BC003766.1 | - | 8 | 1247 | 3491917 | ||
BC090626.1 | - | 8 | 1227 | |||
S81451.1 | - | 7 | 1211 | |||
U57636.1 | - | 6 | 1079 | |||
Y07688.1 | - | 8 | 1527 | |||
Y07689.1 | - | 7 | 1206 |