Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfyb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010914.2 | NP_035044.1 | 5 | 482 | ||
GenBank mRNA | AK010762.1 | - | 5 | 462 | ||
AK011792.1 | - | 5 | 401 | |||
AK079209.1 | - | 5 | 437 | |||
AK146168.1 | - | 5 | 484 | |||
AK154749.1 | - | 5 | 444 | |||
AK155749.1 | - | 5 | 444 | |||
BB858171.1 | - | 5 | 489 | |||
BC010719.1 | - | 5 | 423 | 2811674 | ||
X55316.1 | - | 5 | 478 |