Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ntn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010947.3 | NP_035077.1 | 1 | 709 | ||
GenBank mRNA | AF128866.1 | - | 1 | 549 | ||
AF149094.1 | - | 1 | 345 | |||
AF152418.1 | - | 1 | 464 | |||
AK031199.1 | - | 1 | 854 | |||
AK083904.1 | - | 1 | 728 | |||
AK141856.1 | - | 1 | 620 | |||
BC053038.1 | - | 1 | 832 | |||
BC094362.1 | - | 1 | 709 | 6830466 |