Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CARD9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052813.4 | NP_434700.2 | 7 | 1176 | ||
NM_052814.3 | NP_434701.1 | 7 | 1176 | |||
GenBank mRNA | AF311287.1 | - | 7 | 1137 | ||
AK024001.1 | - | 7 | 1156 | |||
AK127481.1 | - | 2 | 938 | |||
AK292081.1 | - | 7 | 1128 | |||
AK295647.1 | - | 7 | 1015 | |||
BC008877.2 | - | 7 | 1151 | 4139529 |