Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oprl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252565.1 | NP_001239494.1 | 2, 3 | 407 | ||
NM_011012.5 | NP_035142.1 | 2, 3 | 479 | |||
GenBank mRNA | AF043276.1 | - | 3, 4 | 564 | ||
AF043277.1 | - | 2, 3 | 350 | |||
AF043278.1 | - | 2, 3 | 391 | |||
AF062381.1 | - | 1, 2 | 253 | |||
AF075605.1 | - | 1, 2 | 252 | |||
AK031926.1 | - | 3, 4 | 591 | |||
AK038620.1 | - | 2, 3 | 409 | |||
AK043275.1 | - | 3, 4 | 463 | |||
AK044178.1 | - | 3, 4 | 433 | |||
AK079529.1 | - | 2, 3 | 452 | |||
BC050885.1 | - | 2, 3 | 401 | 5703258 | ||
BC051982.1 | - | 2, 3 | 339 | 5695055 | ||
U04952.1 | - | 2, 3 | 400 | |||
U09421.1 | - | 2, 3 | 530 | |||
U14165.1 | - | 1, 2 | 240 | |||
X91813.1 | - | 2, 3 | 410 |
Set Membership: |
G-protein coupled receptor |