Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P2rx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011026.2 | NP_035156.2 | 2 | 313 | ||
GenBank mRNA | AF089751.1 | - | 2 | 247 | ||
AF089752.1 | - | 2 | 247 | |||
AJ251459.1 | - | 2 | 227 | |||
AJ251460.1 | - | 2 | 227 | |||
AJ251461.1 | - | 2 | 227 | |||
AJ251462.1 | - | 2 | 227 | |||
AK150351.1 | - | 2 | 303 | |||
AK162820.1 | - | 2 | 279 | |||
AK163104.1 | - | 2 | 301 | |||
BC002099.1 | - | 2 | 286 | |||
BC005597.1 | - | 2 | 261 | 3482458 | ||
BY150918.1 | - | 2 | 327 | |||
JX417451.1 | - | 2 | 273 |