Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdcd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168491.1 | NP_001161963.1 | 3 | 430 | ||
NM_001168492.1 | NP_001161964.1 | 3 | 430 | |||
NM_011050.4 | NP_035180.2 | 3 | 518 | |||
GenBank mRNA | AK134366.1 | - | 3 | 519 | ||
AK172654.1 | - | 3 | 386 | |||
BC055739.1 | - | 3 | 365 | 6414089 | ||
CT010188.1 | - | 2 | 228 | |||
D50465.1 | - | 2 | 298 | |||
D86344.1 | - | 3 | 314 |