Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pkm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253883.1 | NP_001240812.1 | 3 | 489 | ||
NM_011099.3 | NP_035229.2 | 3 | 489 | |||
GenBank mRNA | AK083076.1 | - | 3 | 282 | ||
AK135397.1 | - | 3 | 282 | |||
AK151724.1 | - | 3 | 355 | |||
AK153483.1 | - | 3 | 355 | |||
AK154695.1 | - | 3 | 282 | |||
AK155110.1 | - | 3 | 282 | |||
AK155655.1 | - | 3 | 285 | |||
AK168943.1 | - | 3 | 282 | |||
AK169872.1 | - | 3 | 261 | |||
AK170892.1 | - | 3 | 285 | |||
AK171023.1 | - | 3 | 282 | |||
AK171033.1 | - | 3 | 285 | |||
AK171106.1 | - | 3 | 286 | |||
AK183643.1 | - | 2 | 68 | |||
AK189659.1 | - | 2 | 68 | |||
AK197420.1 | - | 68 | ||||
AK207130.1 | - | 2 | 68 | |||
AK207766.1 | - | 2 | 68 | |||
AK207924.1 | - | 2 | 68 | |||
AK208026.1 | - | 2 | 68 | |||
AK208591.1 | - | 2 | 68 | |||
AK209065.1 | - | 2 | 68 | |||
AK209125.1 | - | 2 | 68 | |||
AK209411.1 | - | 2 | 68 | |||
AK209972.1 | - | 2 | 68 | |||
AK210823.1 | - | 2 | 68 | |||
AK211460.1 | - | 2 | 69 | |||
AK212924.1 | - | 2 | 68 | |||
AK213742.1 | - | 2 | 68 | |||
AK216406.1 | - | 2 | 68 | |||
AK216783.1 | - | 2 | 68 | |||
AK217172.1 | - | 2 | 68 | |||
AK217516.1 | - | 2 | 68 | |||
BC016619.1 | - | 3 | 249 | 3598842 | ||
BC094663.1 | - | 3 | 228 | 6829532 | ||
D38379.1 | - | 3 | 223 | |||
X97047.1 | - | 2 | 200 |