Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pkig | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039390.2 | NP_001034479.1 | 4, 5 | 448 | ||
NM_001039391.2 | NP_001034480.1 | 3, 4 | 362 | |||
NM_001164053.1 | NP_001157525.1 | 3, 4 | 457 | |||
NM_001164055.1 | NP_001157527.1 | 2, 3 | 292 | |||
NM_011106.2 | NP_035236.1 | 3, 4 | 375 | |||
GenBank mRNA | AK164716.1 | - | 3, 4 | 350 | ||
AK168614.1 | - | 3, 4 | 300 | |||
AK186927.1 | - | 2, 3 | 286 | |||
AK209509.1 | - | 1, 2 | 117 | |||
BC026550.1 | - | 3, 4 | 360 | 5006826 | ||
BY240874.1 | - | 3, 4 | 427 | |||
BY283174.1 | - | 4, 5 | 383 | |||
BY343638.1 | - | 3, 4 | 339 | |||
U97170.1 | - | 3, 4 | 379 | |||
W75470.1 | - | 3, 4 | 388 |