Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pld3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011116.1 | NP_035246.1 | 5, 6 | 515 | ||
GenBank mRNA | AF026124.1 | - | 5, 6 | 515 | ||
AK153508.1 | - | 3, 4 | 204 | |||
BC068170.1 | - | 5, 6 | 512 | |||
BC076586.1 | - | 5, 6 | 579 | 30604186 |