Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P2rx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011026.2 | NP_035156.2 | 2, 3 | 387 | ||
GenBank mRNA | AF089751.1 | - | 2, 3 | 321 | ||
AF089752.1 | - | 2, 3 | 321 | |||
AJ251459.1 | - | 2, 3 | 301 | |||
AJ251460.1 | - | 2, 3 | 301 | |||
AJ251461.1 | - | 2, 3 | 301 | |||
AJ251462.1 | - | 2, 3 | 301 | |||
AK150351.1 | - | 2, 3 | 377 | |||
AK162820.1 | - | 2, 3 | 353 | |||
AK163104.1 | - | 2, 3 | 375 | |||
BC002099.1 | - | 2, 3 | 360 | |||
BC005597.1 | - | 2, 3 | 335 | 3482458 | ||
JX417451.1 | - | 2, 3 | 347 |