Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P2rx7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038839.2 | NP_001033928.1 | 2 | 381 | ||
NM_001038845.2 | NP_001033934.1 | 2 | 381 | |||
NM_001038887.1 | NP_001033976.1 | 2 | 381 | |||
NM_001284402.1 | NP_001271331.1 | 2 | 209 | |||
NM_011027.3 | NP_035157.2 | 2 | 381 | |||
GenBank mRNA | AJ009823.1 | - | 2 | 229 | ||
AJ489296.1 | - | 2 | 218 | |||
AJ489297.1 | - | 2 | 218 | |||
AK144585.1 | - | 2 | 382 | |||
AK145050.1 | - | 2 | 236 | |||
AK145211.1 | - | 2 | 245 | |||
AK171132.1 | - | 2 | 353 | |||
BC062808.1 | - | 2 | 283 | |||
BC128303.1 | - | 2 | 225 | |||
BC128304.1 | - | 2 | 225 | |||
BC141120.1 | - | 2 | 248 | 9055732 | ||
BY752747.1 | - | 2 | 323 | |||
FJ436444.1 | - | 2 | 209 |