Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P4ha2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136076.2 | NP_001129548.1 | 4 | 391 | ||
NM_011031.2 | NP_035161.2 | 4 | 391 | |||
GenBank mRNA | AK053504.1 | - | 4 | 352 | ||
AK142624.1 | - | 4 | 340 | |||
AK144124.1 | - | 4 | 337 | |||
AK145535.1 | - | 4 | 318 | |||
BC018411.1 | - | 4 | 303 | 3157481 | ||
U16163.1 | - | 3 | 278 |