Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdcd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011051.3 | NP_035181.1 | 3, 4 | 324 | ||
GenBank mRNA | AK008610.1 | - | 3, 4 | 248 | ||
AK028223.1 | - | 3, 4 | 279 | |||
AK078044.1 | - | 3, 4 | 252 | |||
AK147979.1 | - | 3, 4 | 282 | |||
AK150335.1 | - | 3, 4 | 242 | |||
BC040079.1 | - | 3, 4 | 275 | 3156800 | ||
BU604816.1 | - | 3, 4 | 327 | |||
U49112.1 | - | 3, 4 | 261 | |||
X14938.1 | - | 2, 3 | 91 |