Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pik3c2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011083.2 | NP_035213.2 | 2 | 952 | ||
GenBank mRNA | AK049609.1 | - | 2 | 942 | ||
AK140908.1 | - | 3 | 1032 | |||
AK142284.1 | - | 3 | 981 | |||
AK163446.1 | - | 2 | 934 | |||
BC096553.1 | - | 1 | 386 | |||
U52193.1 | - | 2 | 942 | |||
U55772.1 | - | 1 | 268 |