Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkacb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164198.1 | NP_001157670.1 | 3, 4 | 504 | ||
NM_001164199.1 | NP_001157671.1 | 3, 4 | 377 | |||
NM_001164200.1 | NP_001157672.1 | 3, 4 | 469 | |||
NM_011100.4 | NP_035230.1 | 3, 4 | 508 | |||
GenBank mRNA | AK048319.1 | - | 3, 4 | 500 | ||
AK139419.1 | - | 3, 4 | 249 | |||
AK148728.1 | - | 3, 4 | 318 | |||
AK150122.1 | - | 3, 4 | 470 | |||
AK150134.1 | - | 3, 4 | 470 | |||
AK163585.1 | - | 3, 4 | 282 | |||
AK193506.1 | - | 3, 4 | 251 | |||
AK204631.1 | - | 3, 4 | 251 | |||
AK216103.1 | - | 3, 4 | 303 | |||
BC054533.1 | - | 3, 4 | 492 | 5704796 | ||
CB723114.1 | - | 3, 4 | 504 | |||
J02626.1 | - | 3, 4 | 437 | |||
JN189786.1 | - | 3, 4 | 320 | |||
JN189787.1 | - | 3, 4 | 269 | |||
JN189788.1 | - | 3, 4 | 244 |