Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OSBPL1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242508.1 | NP_001229437.1 | 16 | 1979 | ||
NM_018030.4 | NP_060500.3 | 14 | 2348 | |||
NM_080597.3 | NP_542164.2 | 28 | 3235 | |||
XM_006722380.3 | XP_006722443.1 | 27 | 3148 | |||
XM_006722382.3 | XP_006722445.1 | 27 | 3113 | |||
XM_017025530.1 | XP_016881019.1 | 28 | 3527 | |||
XM_017025531.1 | XP_016881020.1 | 26 | 2837 | |||
XM_017025533.1 | XP_016881022.1 | 14 | 1831 | |||
XM_017025534.1 | XP_016881023.1 | 14 | 4359 | |||
GenBank mRNA | AF274714.1 | - | 14 | 2348 | ||
AF323726.2 | - | 28 | 3122 | |||
AF392449.1 | - | 14 | 1969 | |||
AF392450.1 | - | 28 | 3191 | |||
BC007004.1 | - | 2 | 346 | |||
BC063420.1 | - | 14 | 2034 | |||
BC098384.1 | - | 14 | 2026 | |||
BC136452.1 | - | 28 | 3132 | 9020441 | ||
BC144117.1 | - | 28 | 3131 | 9052637 |