Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oprk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204371.1 | NP_001191300.1 | 4 | 1346 | ||
NM_011011.2 | NP_035141.1 | 4 | 1316 | |||
GenBank mRNA | AK136866.1 | - | 1 | 240 | ||
AK138198.1 | - | 2 | 2947 | |||
BC116795.1 | - | 4 | 1252 | 40126114 | ||
BC119026.1 | - | 4 | 1252 | 8733775 | ||
BC144731.1 | - | 4 | 1283 | |||
L11065.1 | - | 4 | 1316 | |||
S81111.1 | - | 4 | 1227 |
Set Membership: |
G-protein coupled receptor |