Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oprl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252565.1 | NP_001239494.1 | 4 | 2344 | ||
NM_011012.5 | NP_035142.1 | 4 | 2416 | |||
GenBank mRNA | AF043276.1 | - | 6 | 2383 | ||
AF043278.1 | - | 4 | 2327 | |||
AK031926.1 | - | 5 | 2528 | |||
AK038620.1 | - | 4 | 2346 | |||
AK043275.1 | - | 5 | 2400 | |||
AK044178.1 | - | 5 | 2370 | |||
AK079529.1 | - | 4 | 2389 | |||
AK142474.1 | - | 2 | 1726 | |||
BC050885.1 | - | 4 | 2338 | 5703258 | ||
BC051982.1 | - | 4 | 2276 | 5695055 | ||
U09421.1 | - | 5 | 2349 |
Set Membership: |
G-protein coupled receptor |