Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P2rx7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038839.2 | NP_001033928.1 | 9 | 1071 | ||
NM_001038845.2 | NP_001033934.1 | 9 | 1071 | |||
NM_001284402.1 | NP_001271331.1 | 9 | 899 | |||
NM_011027.3 | NP_035157.2 | 9 | 1071 | |||
GenBank mRNA | AJ009823.1 | - | 9 | 919 | ||
AJ489296.1 | - | 9 | 908 | |||
AJ489297.1 | - | 9 | 908 | |||
AK144585.1 | - | 9 | 1072 | |||
AK171132.1 | - | 9 | 1043 | |||
BC128303.1 | - | 9 | 915 | |||
BC128304.1 | - | 9 | 915 | |||
BC141120.1 | - | 9 | 938 | 9055732 | ||
FJ436444.1 | - | 9 | 899 |