Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcna | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011045.2 | NP_035175.1 | 4 | 545 | ||
GenBank mRNA | AK012614.2 | - | 4 | 547 | ||
AK012730.1 | - | 4 | 546 | |||
AK012932.1 | - | 4 | 545 | |||
AK088256.1 | - | 4 | 546 | |||
AK151238.1 | - | 4 | 546 | |||
AK151411.1 | - | 4 | 578 | |||
AK165217.1 | - | 4 | 546 | |||
AK168233.1 | - | 4 | 546 | |||
AK168596.1 | - | 4 | 549 | |||
BC005778.1 | - | 4 | 530 | 3602797 | ||
BC010343.1 | - | 4 | 529 | 3602977 | ||
X53068.1 | - | 4 | 544 |