Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011049.4 | NP_035179.1 | 16 | 2786 | ||
GenBank mRNA | AK051425.1 | - | 16 | 2787 | ||
AK141083.1 | - | 10 | 1806 | |||
AK145483.1 | - | 16 | 2816 | |||
AK177013.1 | - | 1 | 312 | |||
AK212751.1 | - | 1 | 208 | |||
BC011069.1 | - | 16 | 2545 | 4006745 | ||
X69025.1 | - | 16 | 2808 |